Assay Details
| Gene Symbol: HEXA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261529.1 | NP_001248458.1 | 482 | 80 | ||
| XM_015142597.2 | XP_014998083.1 | 795 | 80 | |||
| GenBank mRNA | JU322568.1 | - | 386 | 80 | ||
| JV046899.1 | - | 482 | 80 | |||
| JV504617.1 | - | 297 | 80 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HEXA
Genomic Map