Assay Details
| Gene Symbol: DENR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151759.2 | XP_015007245.1 | 298 | 157 | ||
| GenBank mRNA | JU336854.1 | - | 155 | 157 | ||
| JV048540.1 | - | 254 | 157 | |||
| JV561315.1 | - | 164 | 157 | |||
| JV699646.1 | - | 249 | 157 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DENR
Genomic Map