Assay Details
| Gene Symbol: MAGED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257614.1 | NP_001244543.1 | 638 | 68 | ||
| XM_015127466.1 | XP_014982952.1 | 765 | 68 | |||
| XM_015127467.2 | XP_014982953.1 | 686 | 68 | |||
| XM_015127468.2 | XP_014982954.1 | 711 | 68 | |||
| XM_015127469.2 | XP_014982955.1 | 762 | 68 | |||
| GenBank mRNA | JU321428.1 | - | 708 | 68 | ||
| JU321429.1 | - | 638 | 68 | |||
| JU471522.1 | - | 699 | 68 | |||
| JU471523.1 | - | 638 | 68 | |||
| JV528057.1 | - | 639 | 68 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MAGED2
Genomic Map