Assay Details
| Gene Symbol: CPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092256.4 | XP_001092256.2 | 1358 | 73 | ||
| XM_015145726.2 | XP_015001212.2 | 1314 | 73 | |||
| XM_015145727.2 | XP_015001213.2 | 1312 | 73 | |||
| XM_028852784.1 | XP_028708617.1 | 1113 | 73 | |||
| XM_028852785.1 | XP_028708618.1 | 1312 | 73 | |||
| GenBank mRNA | JU332499.1 | - | 1083 | 73 | ||
| JV045900.1 | - | 1095 | 73 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CPQ
Genomic Map