Assay Details
| Gene Symbol: CD59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261705.1 | NP_001248634.1 | 179 | 75 | ||
| XM_015114496.2 | XP_014969982.1 | 224 | 75 | |||
| GenBank mRNA | JU332208.1 | - | 171 | 75 | ||
| JU470462.1 | - | 154 | 75 | |||
| JU470463.1 | - | 168 | 75 | |||
| JV044622.1 | - | 179 | 75 | |||
| JV567551.1 | - | 174 | 75 | |||
More Information
Set Membership: |
|

CD59
Genomic Map