Assay Details
| Gene Symbol: FYB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084072.4 | XP_001084072.2 | 1632 | 122 | ||
| XM_001084428.4 | XP_001084428.1 | 1766 | 122 | |||
| XM_015139942.2 | XP_014995428.1 | 1766 | 122 | |||
| XM_015139943.2 | XP_014995429.1 | 1632 | 122 | |||
| XM_028849414.1 | XP_028705247.1 | 1632 | 122 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FYB1
Genomic Map