Assay Details
| Gene Symbol: CCNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849892.1 | XP_028705725.1 | 831 | 89 | ||
| GenBank mRNA | JU321587.1 | - | 812 | 89 | ||
| JU477709.1 | - | 725 | 89 | |||
| JV601125.1 | - | 813 | 89 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CCNG1
Genomic Map