Assay Details
| Gene Symbol: PSMB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260636.1 | NP_001247565.1 | 513 | 94 | ||
| GenBank mRNA | JU322558.1 | - | 513 | 94 | ||
| JV045703.1 | - | 506 | 94 | |||
| JV690223.1 | - | 515 | 94 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PSMB1
Genomic Map