Assay Details
| Gene Symbol: CDCA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086843.3 | XP_001086843.1 | 753 | 128 | ||
| XM_015110388.2 | XP_014965874.1 | 534 | 128 | |||
| GenBank mRNA | JU472138.1 | - | 419 | 128 | ||
| JU472139.1 | - | 656 | 128 | |||
| JU472140.1 | - | 419 | 128 | |||
| JV635460.1 | - | 750 | 128 | |||
| JV636962.1 | - | 750 | 128 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CDCA7
Genomic Map