Assay Details
| Gene Symbol: ANKRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257442.1 | NP_001244371.1 | 404 | 69 | ||
| XM_015121338.2 | XP_014976824.1 | 616 | 69 | |||
| XM_028837092.1 | XP_028692925.1 | 616 | 69 | |||
| GenBank mRNA | JU335362.1 | - | 404 | 69 | ||
| JU470542.1 | - | 484 | 69 | |||
| JU476315.1 | - | 484 | 69 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ANKRD10
Genomic Map