Assay Details
| Gene Symbol: HSD17B10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261171.1 | NP_001248100.1 | 377 | 76 | ||
| GenBank mRNA | JU322857.1 | - | 377 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSD17B10| Gene Symbol: HSD17B10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261171.1 | NP_001248100.1 | 377 | 76 | ||
| GenBank mRNA | JU322857.1 | - | 377 | 76 | ||
Set Membership: |
|