Assay Details
| Gene Symbol: LOC698227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123918.2 | XP_014979404.1 | 3048 | 85 | ||
| GenBank mRNA | FJ613343.1 | - | 251 | 85 | ||
| FJ613344.1 | - | 251 | 85 | |||
| JV513468.1 | - | 210 | 85 | |||
More Information
Set Membership: |
|

LOC698227
Genomic Map