Assay Details
| Gene Symbol: TXNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261708.2 | NP_001248637.1 | 450 | 155 | ||
| XM_015122056.2 | XP_014977542.1 | 454 | 155 | |||
| GenBank mRNA | JU319826.1 | - | 400 | 155 | ||
| JU471046.1 | - | 355 | 155 | |||
| JV044198.1 | - | 450 | 155 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TXNL1
Genomic Map