Assay Details
| Gene Symbol: DGCR6L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193690.2 | NP_001180619.2 | 609 | 72 | ||
| XM_028827116.1 | XP_028682949.1 | 680 | 72 | |||
| GenBank mRNA | JU322664.1 | - | 587 | 72 | ||
| JU477572.1 | - | 589 | 72 | |||
| JV565811.1 | - | 588 | 72 | |||
| Gene Symbol: DGCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149639.2 | XP_015005125.2 | 658 | 72 | ||
| GenBank mRNA | JV044822.1 | - | 577 | 72 | ||
| JV664066.1 | - | 416 | 72 | |||
| JV667290.1 | - | 568 | 72 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DGCR6L
Genomic Map