Assay Details
Gene Symbol: GFUS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261649.1 | NP_001248578.1 | 493 | 98 | ||
GenBank mRNA | JU336193.1 | - | 467 | 98 | ||
JU477039.1 | - | 436 | 98 | |||
JV043366.1 | - | 493 | 98 | |||
JV695119.1 | - | 436 | 98 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Panther Classification:
