Assay Details
| Gene Symbol: MRPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138465.1 | XP_014993951.1 | 1181 | 112 | ||
| GenBank mRNA | JU347353.1 | - | 958 | 112 | ||
| JV068990.1 | - | 855 | 112 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MRPL1
Genomic Map