Assay Details
| Gene Symbol: GDE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260575.1 | NP_001247504.1 | 870 | 129 | ||
| XM_015125655.2 | XP_014981141.1 | 937 | 129 | |||
| XM_028840361.1 | XP_028696194.1 | 1026 | 129 | |||
| GenBank mRNA | JU334931.1 | - | 974 | 129 | ||
| JU471360.1 | - | 974 | 129 | |||
| JV043289.1 | - | 870 | 129 | |||
| JV653455.1 | - | 977 | 129 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GDE1
Genomic Map