Assay Details
| Gene Symbol: DAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257566.1 | NP_001244495.1 | 318 | 64 | ||
| GenBank mRNA | JU333175.1 | - | 318 | 64 | ||
| JU474486.1 | - | 318 | 64 | |||
| JV046303.1 | - | 325 | 64 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DAP
Genomic Map