Assay Details
| Gene Symbol: LOC694684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085895.4 | XP_001085895.1 | 980 | 85 | ||
| XM_028831637.1 | XP_028687470.1 | 524 | 85 | |||
More Information
Set Membership: |
|

LOC694684
Genomic Map