Assay Details
| Gene Symbol: NARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122067.2 | XP_014977553.2 | 250 | 117 | ||
| GenBank mRNA | JV469498.1 | - | 146 | 117 | ||
| JV617559.1 | - | 282 | 117 | |||
More Information
Set Membership: |
|

NARS1
Genomic Map