Assay Details
| Gene Symbol: USP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085998.4 | XP_001085998.3 | 1331 | 94 | ||
| XM_015141078.1 | XP_014996564.1 | 483 | 94 | |||
| GenBank mRNA | JU322805.1 | - | 917 | 94 | ||
| JU471509.1 | - | 878 | 94 | |||
| JV044688.1 | - | 694 | 94 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

USP1
Genomic Map