Assay Details
Gene Symbol: MGARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193816.2 | NP_001180745.2 | 467 | 120 | ||
GenBank mRNA | JV725365.1 | - | 180 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
