Assay Details
| Gene Symbol: CDIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194838.2 | NP_001181767.1 | 575 | 133 | ||
| XM_001097426.4 | XP_001097426.1 | 854 | 133 | |||
| XM_001097525.3 | XP_001097525.1 | 719 | 133 | |||
| XM_028840479.1 | XP_028696312.1 | 717 | 133 | |||
| XM_028840480.1 | XP_028696313.1 | 798 | 133 | |||
| XM_028840481.1 | XP_028696314.1 | 710 | 133 | |||
| GenBank mRNA | JU321653.1 | - | 716 | 133 | ||
| JU471490.1 | - | 702 | 133 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CDIP1
Genomic Map