Assay Details
| Gene Symbol: PBDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194106.1 | NP_001181035.1 | 227 | 56 | ||
| XM_015127705.2 | XP_014983191.1 | 302 | 56 | |||
| XM_028841782.1 | XP_028697615.1 | 298 | 56 | |||
| GenBank mRNA | JU322812.1 | - | 127 | 56 | ||
| JU475866.1 | - | 127 | 56 | |||
| JV508990.1 | - | 130 | 56 | |||
| JV653154.1 | - | 149 | 56 | |||
More Information
Set Membership: |
|

PBDC1
Genomic Map