Assay Details
| Gene Symbol: KBTBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260755.1 | NP_001247684.1 | 111 | 69 | ||
| XM_015115577.2 | XP_014971063.2 | 462 | 69 | |||
| GenBank mRNA | JU333047.1 | - | 111 | 69 | ||
| JV707425.1 | - | 34 | 69 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260755.1, XM_015115577.2 transcripts.
Set Membership: |
|

KBTBD3
Genomic Map