Assay Details
| Gene Symbol: CA5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101899.4 | XP_001101899.1 | 588 | 84 | ||
| GenBank mRNA | JV045715.1 | - | 578 | 84 | ||
| JV635611.1 | - | 538 | 84 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CA5B
Genomic Map