Assay Details
| Gene Symbol: EIF4E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266219.2 | NP_001253148.1 | 140 | 110 | ||
| GenBank mRNA | JU320282.1 | - | 152 | 110 | ||
| JU470709.1 | - | 147 | 110 | |||
| JV044801.1 | - | 143 | 110 | |||
| JV675335.1 | - | 129 | 110 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EIF4E
Genomic Map