Assay Details
| Gene Symbol: BPGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266072.2 | NP_001253001.1 | 108 | 114 | ||
| GenBank mRNA | JV048024.1 | - | 109 | 114 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the NM_001266072.2 transcripts.
This assay is in the 5'UTR region of the NM_001266072.2 transcripts.
Set Membership: |
|

BPGM
Genomic Map