Assay Details
| Gene Symbol: C1QA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130417.1 | XP_014985903.1 | 191 | 97 | ||
| GenBank mRNA | JV480266.1 | - | 78 | 97 | ||
| JV722280.1 | - | 78 | 97 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015130417.1 transcripts.
Set Membership: |
|

C1QA
Genomic Map