Assay Details
| Gene Symbol: CD200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257544.1 | NP_001244473.1 | 200 | 94 | ||
| XM_015129804.2 | XP_014985290.1 | 338 | 94 | |||
| XM_028843273.1 | XP_028699106.1 | 395 | 94 | |||
| XM_028843277.1 | XP_028699110.1 | 341 | 94 | |||
| XM_028843279.1 | XP_028699112.1 | 338 | 94 | |||
| GenBank mRNA | JU323326.1 | - | 200 | 94 | ||
| JU470812.1 | - | 215 | 94 | |||
| JV044869.1 | - | 132 | 94 | |||
| JV715409.1 | - | 201 | 94 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_028843277.1, XM_028843279.1 transcripts.
This assay is in the 5'UTR region of the XM_028843277.1, XM_028843279.1 transcripts.
Set Membership: |
|

CD200
Genomic Map