Assay Details
| Gene Symbol: RNF138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100574.4 | XP_001100574.1 | 739 | 155 | ||
| XM_015121784.2 | XP_014977270.1 | 552 | 155 | |||
| GenBank mRNA | JU326834.1 | - | 373 | 155 | ||
| JU326835.1 | - | 373 | 155 | |||
| JU478331.1 | - | 415 | 155 | |||
| JV716587.1 | - | 373 | 155 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RNF138
Genomic Map