Assay Details
| Gene Symbol: CHMP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266802.1 | NP_001253731.1 | 527 | 70 | ||
| XM_028843445.1 | XP_028699278.1 | 637 | 70 | |||
| GenBank mRNA | JU333892.1 | - | 575 | 70 | ||
| JU471964.1 | - | 522 | 70 | |||
| JV046696.1 | - | 527 | 70 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CHMP2B
Genomic Map