Assay Details
| Gene Symbol: PAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265935.1 | NP_001252864.1 | 1431 | 142 | ||
| GenBank mRNA | JU321671.1 | - | 1604 | 142 | ||
| JU471801.1 | - | 1655 | 142 | |||
| JV044277.1 | - | 1431 | 142 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PAK2
Genomic Map