Assay Details
| Gene Symbol: PXMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149188.2 | XP_015004674.1 | 330 | 97 | ||
| GenBank mRNA | JU323044.1 | - | 181 | 97 | ||
| JV455580.1 | - | 84 | 97 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PXMP4
Genomic Map