Assay Details
| Gene Symbol: RACK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105066.4 | XP_001105066.1 | 1176 | 89 | ||
| GenBank mRNA | JV670477.1 | - | 865 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RACK1| Gene Symbol: RACK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105066.4 | XP_001105066.1 | 1176 | 89 | ||
| GenBank mRNA | JV670477.1 | - | 865 | 89 | ||
Set Membership: |
|