Assay Details
| Gene Symbol: ALDH3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118800.2 | XP_014974286.1 | 1139 | 79 | ||
| XM_015118801.2 | XP_014974287.1 | 788 | 79 | |||
| GenBank mRNA | JU322743.1 | - | 716 | 79 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ALDH3A1
Genomic Map