Assay Details
| Gene Symbol: LOC714004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001103553.4 | XP_001103553.2 | 324 | 118 | ||
| XM_015144127.2 | XP_014999613.1 | 425 | 118 | |||
| XM_028831953.1 | XP_028687786.1 | 325 | 118 | |||
| XM_028831955.1 | XP_028687788.1 | 325 | 118 | |||
More Information
Set Membership: |
|

LOC714004
Genomic Map