Assay Details
| Gene Symbol: UBD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001102785.4 | XP_001102785.2 | 183 | 78 | ||
| GenBank mRNA | JU474005.1 | - | 53 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001102785.4 transcripts.
Set Membership: |
|

UBD
Genomic Map