Assay Details
| Gene Symbol: TOMM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265860.1 | NP_001252789.1 | 129 | 97 | ||
| GenBank mRNA | JU472350.1 | - | 129 | 97 | ||
| JV044978.1 | - | 129 | 97 | |||
More Information
Set Membership: |
|

TOMM7
Genomic Map