Assay Details
| Gene Symbol: TOMM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265860.1 | NP_001252789.1 | 180 | 125 | ||
| GenBank mRNA | JU472350.1 | - | 180 | 125 | ||
| JV044978.1 | - | 180 | 125 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TOMM7
Genomic Map