Assay Details
| Gene Symbol: MRPS27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098829.4 | XP_001098829.2 | 728 | 92 | ||
| XM_015140263.2 | XP_014995749.2 | 669 | 92 | |||
| XM_028849504.1 | XP_028705337.1 | 716 | 92 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MRPS27
Genomic Map