Assay Details
| Gene Symbol: SPARC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141267.2 | XP_014996753.1 | 420 | 122 | ||
| XM_028849854.1 | XP_028705687.1 | 444 | 122 | |||
| XM_028849855.1 | XP_028705688.1 | 532 | 122 | |||
| GenBank mRNA | JU336059.1 | - | 442 | 122 | ||
| JU476920.1 | - | 402 | 122 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SPARC
Genomic Map