Assay Details
| Gene Symbol: RNF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266146.1 | NP_001253075.1 | 369 | 71 | ||
| GenBank mRNA | JU320228.1 | - | 386 | 71 | ||
| JU475167.1 | - | 342 | 71 | |||
| JV044695.1 | - | 369 | 71 | |||
| JV602193.1 | - | 605 | 71 | |||
| JV667573.1 | - | 592 | 71 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RNF11
Genomic Map