Assay Details
| Gene Symbol: SEPTIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111558.2 | XP_014967044.1 | 733 | 124 | ||
| XM_015111559.2 | XP_014967045.1 | 809 | 124 | |||
| XM_028831357.1 | XP_028687190.1 | 723 | 124 | |||
| XM_028831358.1 | XP_028687191.1 | 711 | 124 | |||
| XM_028831359.1 | XP_028687192.1 | 738 | 124 | |||
| GenBank mRNA | JV047156.1 | - | 915 | 124 | ||
| JV636795.1 | - | 881 | 124 | |||
| Gene Symbol: HDLBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU470468.1 | - | 1267 | 124 | ||
| JV043383.1 | - | 1262 | 124 | |||
| JV649682.1 | - | 1266 | 124 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SEPTIN2
Genomic Map