Assay Details
| Gene Symbol: NDUFAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112115.2 | XP_014967601.2 | 505 | 118 | ||
| XM_015112116.2 | XP_014967602.2 | 371 | 118 | |||
| XM_015112117.2 | XP_014967603.2 | 488 | 118 | |||
| XM_015112118.2 | XP_014967604.2 | 437 | 118 | |||
| GenBank mRNA | JU335336.1 | - | 454 | 118 | ||
| JU476289.1 | - | 476 | 118 | |||
| JV047570.1 | - | 420 | 118 | |||
| JV655459.1 | - | 481 | 118 | |||
| JV718271.1 | - | 454 | 118 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NDUFAF7
Genomic Map