Assay Details
Gene Symbol: RAMAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265849.1 | NP_001252778.1 | 357 | 92 | ||
GenBank mRNA | JU321796.1 | - | 357 | 92 | ||
JU472742.1 | - | 360 | 92 | |||
JV043700.1 | - | 357 | 92 | |||
JV638339.1 | - | 362 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
