Assay Details
Gene Symbol: SAR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261028.1 | NP_001247957.1 | 345 | 94 | ||
XM_015140863.2 | XP_014996349.1 | 90 | 94 | |||
GenBank mRNA | JU322264.1 | - | 371 | 94 | ||
JU322265.1 | - | 371 | 94 | |||
JU322266.1 | - | 371 | 94 | |||
JU322267.1 | - | 371 | 94 | |||
JU473494.1 | - | 362 | 94 | |||
JV044122.1 | - | 345 | 94 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
