Assay Details
Gene Symbol: GAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107824.4 | XP_001107824.1 | 1039 | 61 | ||
XM_028843446.1 | XP_028699279.1 | 692 | 61 | |||
GenBank mRNA | JU322581.1 | - | 742 | 61 | ||
JV452944.1 | - | 916 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
