Assay Details
| Gene Symbol: VPS29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107808.4 | XP_001107808.1 | 352 | 97 | ||
| XM_001107873.3 | XP_001107873.1 | 298 | 97 | |||
| XM_015152780.2 | XP_015008266.1 | 288 | 97 | |||
| GenBank mRNA | JU328140.1 | - | 69 | 97 | ||
| JV547659.1 | - | 126 | 97 | |||
| JV686659.1 | - | 189 | 97 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

VPS29
Genomic Map