Assay Details
| Gene Symbol: LOC712203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119807.2 | XP_014975293.1 | 1032 | 83 | ||
| XM_028836531.1 | XP_028692364.1 | 1032 | 83 | |||
| XM_028836532.1 | XP_028692365.1 | 1032 | 83 | |||
| GenBank mRNA | JU348939.1 | - | 755 | 83 | ||
| JV558932.1 | - | 734 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC712203
Genomic Map